Assay Details
| Gene Symbol: RNF145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX537635.1 | - | 1 | 613 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BX537635.1 transcripts.
Set Membership: |
|

RNF145
Genomic Map