| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |