Assay Details
| Gene Symbol: DCLRE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540429.1 | XP_011538731.1 | 1 - 2 | 118 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011540429.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via nonhomologous end joining
mitotic nuclear division
protection from non-homologous end joining at telomere
interstrand cross-link repair
cell division
nucleic acid phosphodiester bond hydrolysis
damaged DNA binding
5'-3' exodeoxyribonuclease activity
double-strand break repair via nonhomologous end joining
mitotic nuclear division
protection from non-homologous end joining at telomere
interstrand cross-link repair
cell division
nucleic acid phosphodiester bond hydrolysis
damaged DNA binding
5'-3' exodeoxyribonuclease activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |

DCLRE1A
Genomic Map