Assay Details
| Gene Symbol: HMGN2P46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096745.1 | - | 2 - 3 | 145 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK096745.1 transcripts.
Set Membership: |
|

HMGN2P46
Genomic Map