Assay Details
| Gene Symbol: HOMER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006720775.3 | XP_006720838.1 | 10 | 7447 | 108 | |
| GenBank mRNA | CR936744.1 | - | 2 | 5866 | 108 | |
More Information
Set Membership: |
|

HOMER2
Genomic Map