Assay Details
Gene Symbol: LOC283728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK095617.1 | - | 1 | 556 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000516761.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | ENST00000620198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007263.hg.1 | ENST00000612245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006799.hg.1 | ENST00000621346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000398377.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000423852.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000516761.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | ENST00000620198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_001555.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007242.hg.1 | NR_002195.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000441780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007263.hg.1 | ENST00000612245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000417252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006799.hg.1 | ENST00000621346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008141.hg.1 | ENST00000561247.1 | Ensembl |
