Assay Details
| Gene Symbol: TTC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012382.2 | NP_036514.1 | 5 | 4427 | 93 | |
| GenBank mRNA | BM543635.1 | - | 434 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC33| Gene Symbol: TTC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012382.2 | NP_036514.1 | 5 | 4427 | 93 | |
| GenBank mRNA | BM543635.1 | - | 434 | 93 | ||
Set Membership: |
|