Assay Details
| Gene Symbol: ANKS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248964.3 | XP_005249021.1 | 4 - 5 | 628 | 56 | |
| XM_006715036.2 | XP_006715099.1 | 4 - 5 | 628 | 56 | ||
| XM_011514431.2 | XP_011512733.1 | 4 - 5 | 628 | 56 | ||
| XM_011514432.2 | XP_011512734.1 | 4 - 5 | 628 | 56 | ||
| XM_011514433.1 | XP_011512735.1 | 4 - 5 | 628 | 56 | ||
| XM_011514436.2 | XP_011512738.1 | 4 - 5 | 4197 | 56 | ||
More Information
Set Membership: |
|

ANKS1A
Genomic Map