Assay Details
| Gene Symbol: AFMID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011524328.2 | XP_011522630.1 | 2 - 3 | 124 | 100 | |
| XM_011524332.2 | XP_011522634.1 | 2 - 3 | 124 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011524328.2, XM_011524332.2 transcripts.
Set Membership: |
|

AFMID
Genomic Map