Assay Details
| Gene Symbol: MAPK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010299.1 | XP_016865788.1 | 11 - 12 | 1432 | 77 | |
| XM_017010300.1 | XP_016865789.1 | 11 - 12 | 1432 | 77 | ||
More Information
Set Membership: |
|

MAPK14
Genomic Map