Assay Details
| Gene Symbol: LOC107986596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011613.1 | XP_016867102.1 | 3 - 4 | 2375 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107986596| Gene Symbol: LOC107986596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011613.1 | XP_016867102.1 | 3 - 4 | 2375 | 110 | |
Set Membership: |
|