Assay Details
| Gene Symbol: CYP3A43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278921.1 | NP_001265850.1 | 9 - 10 | 1191 | 175 | |
| NM_022820.4 | NP_073731.1 | 12 - 13 | 1524 | 175 | ||
| NM_057095.2 | NP_476436.1 | 12 - 13 | 1521 | 175 | ||
| NR_103869.1 | 13 - 14 | 1642 | 175 | |||
| XM_017012542.1 | XP_016868031.1 | 10 - 11 | 1192 | 175 | ||
| XM_017012543.1 | XP_016868032.1 | 9 - 10 | 1350 | 175 | ||
| XM_017012544.1 | XP_016868033.1 | 8 - 9 | 1078 | 175 | ||
| XM_017012545.1 | XP_016868034.1 | 8 - 9 | 1058 | 175 | ||
| GenBank mRNA | AA417369.1 | - | 1 - 2 | 175 | ||
| AF337813.1 | - | 12 - 13 | 1521 | 175 | ||
More Information
Set Membership: |
|

CYP3A43
Genomic Map