Assay Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250108.1 | XP_005250165.1 | 1 - 2 | 145 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005250108.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000249377.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000339431.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000455453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000485478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | ENST00000498487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_001031692.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008657.hg.1 | NM_005824.2 | Refseq |

LRRC17
Genomic Map