| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000490162.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000490162.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000490162.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000490162.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012192.hg.1 |
ENST00000516169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000470135.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000490162.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000490162.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000490162.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000490162.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008741.hg.1 |
ENST00000602761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012192.hg.1 |
ENST00000516169.1 |
Ensembl |