Assay Details
| Gene Symbol: HOXA11-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002795.2 | 2 | 1480 | 110 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000613383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000613939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000615604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000615604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000620092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000620211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | ENST00000620901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000479766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000520395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000522863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000613383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000613939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000615604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000615604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000620092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000620211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | ENST00000620901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007006.hg.1 | NR_002795.2 | Refseq |

HOXA11-AS
Genomic Map