Assay Details
| Gene Symbol: WNT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK294096.1 | - | 1 - 2 | 260 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK294096.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000538854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000538854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000542408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000542408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000542408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000542408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000543563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000310594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000397196.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000537031.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000538854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000538854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000539198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000542408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000542408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000542408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000542408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543071.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000543563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | ENST00000545811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_030775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006469.hg.1 | NM_032642.2 | Refseq |

WNT5B
Genomic Map