Assay Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033017.3 | NP_148977.2 | 3 - 4 | 761 | 91 | |
| NM_033091.2 | NP_149082.1 | 2 - 3 | 683 | 91 | ||
| GenBank mRNA | AF220023.1 | - | 4 - 5 | 698 | 91 | |
| AF220024.1 | - | 2 - 3 | 620 | 91 | ||
| AK092892.1 | - | 2 - 3 | 645 | 91 | ||
| AK293565.1 | - | 4 - 5 | 537 | 91 | ||
| BC011763.2 | - | 2 - 3 | 576 | 3617674 | 91 | |
| BC025949.1 | - | 2 - 3 | 576 | 4620469 | 91 | |
More Information
Set Membership: |
|

TRIM4
Genomic Map