Assay Details
| Gene Symbol: TRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317804.1 | NP_001304733.1 | 12 | 2563 | 92 | |
| NM_001317805.1 | NP_001304734.1 | 11 | 3078 | 92 | ||
| NM_014294.5 | NP_055109.1 | 11 | 2646 | 92 | ||
| XM_017013266.1 | XP_016868755.1 | 10 | 2518 | 92 | ||
| XM_017013267.1 | XP_016868756.1 | 10 | 2862 | 92 | ||
| GenBank mRNA | AF130055.2 | - | 1 | 92 | ||
| AF274933.1 | - | 1 | 765 | 92 | ||
| BC000687.2 | - | 11 | 2468 | 3347823 | 92 | |
| BC037738.1 | - | 11 | 2558 | 5261490 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF130055.2 transcripts.
Set Membership: |
|

TRAM1
Genomic Map