Assay Details
| Gene Symbol: TNFRSF10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC043400.1 | - | 1 - 2 | 207 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF10B| Gene Symbol: TNFRSF10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC043400.1 | - | 1 - 2 | 207 | 108 | |
Set Membership: |
|