Assay Details
| Gene Symbol: EYA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035138.1 | - | 3 - 4 | 331 | 59 | |
| BC048982.1 | - | 3 - 4 | 325 | 59 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000523987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000523987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000523987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012425.hg.1 | ENST00000523987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000519927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000523987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000523987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000523987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012425.hg.1 | ENST00000523987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |

EYA1
Genomic Map