Assay Details
| Gene Symbol: LOC101929709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BQ652687.1 | - | 2 - 3 | 371 | 88 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |

LOC101929709
Genomic Map