Assay Details
| Gene Symbol: CRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000756.3 | NP_000747.1 | 2 | 486 | 107 | |
| XM_017013090.1 | XP_016868579.1 | 6 | 824 | 107 | ||
| XM_017013091.1 | XP_016868580.1 | 6 | 887 | 107 | ||
| XM_017013092.1 | XP_016868581.1 | 6 | 825 | 107 | ||
| XM_017013093.1 | XP_016868582.1 | 6 | 964 | 107 | ||
| GenBank mRNA | AK075431.1 | - | 2 | 432 | 107 | |
| BC002599.2 | - | 2 | 297 | 3160933 | 107 | |
| BC011031.1 | - | 2 | 307 | 4280754 | 107 | |
| BT007453.1 | - | 1 | 119 | 107 | ||
| CB992265.1 | - | 2 | 513 | 107 | ||
| KU177994.1 | - | 1 | 119 | 107 | ||
More Information
Set Membership: |
|

CRH
Genomic Map