Assay Details
| Gene Symbol: MAMDC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124126.1 | - | 2 - 3 | 429 | 98 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000460688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000460688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000460688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | ENST00000460688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000420573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000420573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000377182.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000460688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000460688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000460688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | ENST00000460688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007518.hg.1 | NM_153267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000377178.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000420573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000420573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000448377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000451488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | ENST00000535188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012242.hg.1 | NR_040076.1 | Refseq |

MAMDC2-AS1
Genomic Map