Assay Details
| Gene Symbol: SPTLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281303.1 | NP_001268232.1 | 15 | 115 | ||
| NM_006415.3 | NP_006406.1 | 15 | 115 | |||
| XM_011518139.2 | XP_011516441.1 | 15 | 115 | |||
| XM_017014200.1 | XP_016869689.1 | 16 | 115 | |||
| XM_017014201.1 | XP_016869690.1 | 16 | 115 | |||
| XM_017014202.1 | XP_016869691.1 | 11 | 115 | |||
| GenBank mRNA | AK098344.1 | - | 4 | 115 | ||
| BC068537.1 | - | 15 | 5259635 | 115 | ||
More Information
Set Membership: |
|

SPTLC1
Genomic Map