| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000336949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000378474.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000457894.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
ENST00000614558.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_001098791.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006977.hg.1 |
NM_021242.5 |
Refseq |