| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000266775.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000392872.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000401157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000436021.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000436021.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000436021.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000436021.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000436021.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000436021.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000436021.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000536395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000536395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000536395.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000537100.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000540956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000542926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000544861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000545698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000545698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000545698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000545698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
ENST00000545698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008669.hg.1 |
NM_003211.4 |
Refseq |