Assay Details
| Gene Symbol: CCDC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286243.1 | NP_001273172.1 | 3 - 4 | 501 | 105 | |
| NM_001286244.1 | NP_001273173.1 | 2 - 3 | 331 | 105 | ||
| NM_152591.2 | NP_689804.1 | 4 - 5 | 606 | 105 | ||
| XM_006719262.1 | XP_006719325.1 | 5 - 6 | 798 | 105 | ||
| XM_006719263.2 | XP_006719326.1 | 4 - 5 | 559 | 105 | ||
| XM_011537999.1 | XP_011536301.1 | 5 - 6 | 849 | 105 | ||
| XM_011538001.2 | XP_011536303.1 | 5 - 6 | 841 | 105 | ||
| GenBank mRNA | AK093162.1 | - | 4 - 5 | 559 | 105 | |
| AK302207.1 | - | 3 - 4 | 501 | 105 | ||
| AK310294.1 | - | 2 - 3 | 522 | 105 | ||
| BC044815.1 | - | 4 - 5 | 525 | 105 | ||
| BC064580.1 | - | 2 - 3 | 264 | 105 | ||
| BC073147.1 | - | 3 - 4 | 415 | 5742508 | 105 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |

CCDC63
Genomic Map