Assay Details
| Gene Symbol: CCDC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310294.1 | - | 3 - 4 | 649 | 79 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |

CCDC63
Genomic Map