Assay Details
| Gene Symbol: SOCS2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054607.1 | - | 3 - 4 | 526 | 67 | |
| AK098304.1 | - | 2 - 3 | 430 | 67 | ||
| BC040677.1 | - | 410 | 67 | |||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |

SOCS2-AS1
Genomic Map