Assay Details
| Gene Symbol: LOC100505502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519798.2 | XP_011518100.1 | 1 | 1194 | 99 | |
| GenBank mRNA | AK094743.1 | - | 2 | 1293 | 99 | |
More Information
Set Membership: |
|

LOC100505502
Genomic Map