Assay Details
| Gene Symbol: WAC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033805.1 | 2 - 3 | 677 | 98 | ||
| GenBank mRNA | BC045818.1 | - | 2 - 3 | 657 | 98 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |

WAC-AS1
Genomic Map