| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000228534.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000619177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
ENST00000622119.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007838.hg.1 |
NM_016584.2 |
Refseq |