| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000531285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000531285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
ENST00000531285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000522111.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000531285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000531285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
ENST00000531285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007493.hg.1 |
NM_001013642.2 |
Refseq |