| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000358582.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000585624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000590008.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
ENST00000593040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011936.hg.1 |
NM_152605.3 |
Refseq |