Assay Details
| Gene Symbol: PINLYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193621.1 | NP_001180550.1 | 4 - 5 | 544 | 62 | |
| NM_001193622.1 | NP_001180551.1 | 2 - 3 | 204 | 62 | ||
| NM_001321124.1 | NP_001308053.1 | 2 - 3 | 224 | 62 | ||
| XM_011526970.2 | XP_011525272.1 | 3 - 4 | 2140 | 62 | ||
| XM_011526971.2 | XP_011525273.1 | 2 - 3 | 566 | 62 | ||
| GenBank mRNA | AK129550.1 | - | 2 - 3 | 192 | 62 | |
| BC071811.1 | - | 2 - 3 | 204 | 62 | ||
| CN355575.1 | - | 2 - 3 | 183 | 62 | ||
| DW009999.1 | - | 2 - 3 | 62 | |||
| HY005068.1 | - | 2 - 3 | 224 | 62 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008220.hg.1 | ENST00000600242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008220.hg.1 | ENST00000600242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008220.hg.1 | ENST00000600242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008220.hg.1 | ENST00000600242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000244321.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000413422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000562365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000569031.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000599207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | ENST00000612042.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008221.hg.1 | NM_001193622.1 | Refseq |

PINLYP
Genomic Map