Assay Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303539.1 | NP_001290468.1 | 2 - 3 | 573 | 107 | |
| NM_018273.3 | NP_060743.2 | 2 - 3 | 573 | 107 | ||
| GenBank mRNA | AK129801.1 | - | 2 - 3 | 276 | 107 | |
| AK298317.1 | - | 2 - 3 | 274 | 107 | ||
| AK303635.1 | - | 2 - 3 | 276 | 107 | ||
| BC016919.1 | - | 2 - 3 | 265 | 4400601 | 107 | |
| BC040021.1 | - | 2 - 3 | 271 | 107 | ||
More Information
Set Membership: |
|

TMEM143
Genomic Map