Assay Details
| Gene Symbol: NDUFA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005002.4 | NP_004993.1 | 5 - 6 | 116 | ||
| GenBank mRNA | AF050641.1 | - | 5 - 6 | 116 | ||
| AK291067.1 | - | 5 - 6 | 116 | |||
| AK308588.1 | - | 5 - 6 | 116 | |||
| BC003351.1 | - | 4 - 5 | 116 | |||
| BC009311.2 | - | 5 - 6 | 4106931 | 116 | ||
| BC015837.1 | - | 5 - 6 | 4293690 | 116 | ||
| BC111546.1 | - | 5 - 6 | 5761015 | 116 | ||
| L04490.1 | - | 5 - 6 | 116 | |||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC022214.1 | - | 735 | Chr. - - [-] on Build | 116 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial electron transport, NADH to ubiquinone
sodium ion transport
mitochondrial respiratory chain complex I assembly
ubiquinone-6 biosynthetic process
NADH dehydrogenase activity
protein binding
NADH dehydrogenase (ubiquinone) activity
protein complex binding
coenzyme binding
mitochondrial electron transport, NADH to ubiquinone
sodium ion transport
mitochondrial respiratory chain complex I assembly
ubiquinone-6 biosynthetic process
NADH dehydrogenase activity
protein binding
NADH dehydrogenase (ubiquinone) activity
protein complex binding
coenzyme binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000540688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000540688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000540688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000540688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000542369.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000542369.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000542369.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000542369.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | ENST00000544679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000266544.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000396655.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000535726.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000539573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000540688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000540688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000540688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000540688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000542369.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000542369.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000542369.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000542369.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | ENST00000544679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012576.hg.1 | NM_005002.4 | Refseq |

NDUFA9
Genomic Map