| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008136.hg.1 |
ENST00000414992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008136.hg.1 |
ENST00000414992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008136.hg.1 |
ENST00000414992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008136.hg.1 |
ENST00000414992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008136.hg.1 |
ENST00000453584.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008136.hg.1 |
ENST00000453584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008136.hg.1 |
ENST00000414992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008136.hg.1 |
ENST00000414992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008136.hg.1 |
ENST00000414992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008136.hg.1 |
ENST00000414992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008136.hg.1 |
ENST00000453584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008136.hg.1 |
ENST00000453584.1 |
Ensembl |