| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000467169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000467169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000467169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000483678.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000483678.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000483678.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000498692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000498692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
ENST00000498692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000375348.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000467169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000467169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000467169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000483678.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000483678.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000483678.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000488362.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000498692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000498692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
ENST00000498692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300010007.hg.1 |
NM_182614.3 |
Refseq |