Assay Details
| Gene Symbol: NUP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020880.1 | XP_016876369.1 | 3 - 4 | 867 | 139 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017020880.1 transcripts.
Set Membership: |
|

NUP58
Genomic Map