Assay Details
| Gene Symbol: ARGLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK309941.1 | - | 1 - 2 | 212 | 79 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |

ARGLU1
Genomic Map