| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000473775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000473775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000473775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000473775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |