| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000550583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000550583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000550583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000550583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000551170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000551170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000551170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000551170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000551871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000551871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000551871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000551871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000552126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000552126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000552126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000552126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000556267.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
ENST00000556267.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000546892.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000548372.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000549809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000550583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000550583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000550583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000550583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000551170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000551170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000551170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000551170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000551871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000551871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000551871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000551871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000552126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000552126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000552126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000552126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000556267.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
ENST00000556267.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010696.hg.1 |
NM_001080464.2 |
Refseq |