Assay Details
| Gene Symbol: AREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011537415.2 | XP_011535717.1 | 2 - 3 | 236 | 103 | |
| XM_011537417.2 | XP_011535719.1 | 2 - 3 | 245 | 103 | ||
| GenBank mRNA | BX248747.1 | - | 2 - 3 | 250 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011537415.2, XM_011537417.2 transcripts.
Set Membership: |
|

AREL1
Genomic Map