Assay Details
| Gene Symbol: IGHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130465.1 | - | 1 - 2 | 83 | 95 | |
| Gene Symbol: IGHG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC075842.1 | - | 1 - 2 | 71 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK130465.1 transcripts.
Set Membership: |
|

IGHA1
Genomic Map