Assay Details
| Gene Symbol: LINC00640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038358.1 | 3 - 4 | 302 | 91 | ||
| GenBank mRNA | BX111479.1 | - | 3 - 4 | 154 | 91 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000555801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000555801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | ENST00000557295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000554409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000555801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000555801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000556479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | ENST00000557295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | NR_038358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007142.hg.1 | NR_038358.1 | Refseq |

LINC00640
Genomic Map