| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
ENST00000549576.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
ENST00000330493.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
ENST00000549576.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011236.hg.1 |
NR_038135.2 |
Refseq |