Assay Details
| Gene Symbol: ANXA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320698.1 | NP_001307627.1 | 1 - 2 | 1096 | 92 | |
| XM_017003943.1 | XP_016859432.1 | 1 - 2 | 801 | 92 | ||
| XM_017003944.1 | XP_016859433.1 | 1 - 2 | 801 | 92 | ||
| GenBank mRNA | DN994496.1 | - | 1 - 2 | 148 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320698.1, XM_017003943.1, XM_017003944.1 transcripts.
Set Membership: |
|

ANXA4
Genomic Map