Assay Details
| Gene Symbol: PRR14L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173566.2 | NP_775837.2 | 2 - 3 | 668 | 66 | |
| XM_006724219.3 | XP_006724282.1 | 2 - 3 | 1228 | 66 | ||
| XM_006724220.3 | XP_006724283.1 | 2 - 3 | 630 | 66 | ||
| XM_011530080.2 | XP_011528382.1 | 3 - 4 | 1436 | 66 | ||
| XM_011530081.2 | XP_011528383.1 | 2 - 3 | 1312 | 66 | ||
| XM_017028736.1 | XP_016884225.1 | 2 - 3 | 2388 | 66 | ||
| XM_017028737.1 | XP_016884226.1 | 1 - 2 | 1826 | 66 | ||
| GenBank mRNA | CT841517.1 | - | 1 - 2 | 500 | 66 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |

PRR14L
Genomic Map