| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000407997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000461827.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000461827.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000461827.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000461827.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000461827.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000461827.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000463934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000463934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000463934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000463934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000463934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000463934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000480000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000480000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000480000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000480000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000480000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000480000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000480000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000481958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000481958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000481958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000481958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000481958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000481958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000481958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000486357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000486357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000486357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000494150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000494150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000494150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000494150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000494150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000494150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000494150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
ENST00000494150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009274.hg.1 |
NM_021822.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000361441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
ENST00000428892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009271.hg.1 |
NM_014508.2 |
Refseq |